ENA413165363 Enamine

4-[butyl(methyl)amino]pyridine-2-carbonitrile

  • MDL number MFCD17242190
  • Cas Number : 1249999-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136827

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136827
Availability details for
Quantity
  • Availability
  • Add to Cart