ENA413165373 Enamine

2-(6-fluoro-1-methyl-1H-indol-3-yl)acetonitrile

  • MDL number MFCD21984807
  • Cas Number : 1368450-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64...66
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137061

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137061
Availability details for
Quantity
  • Availability
  • Add to Cart