ENA413165387 Enamine

4-[3-(trifluoromethyl)phenyl]butanenitrile

  • MDL number MFCD09744586
  • Cas Number : 1057676-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137366
Availability details for
Quantity
  • Availability
  • Add to Cart