ENA413165389 Enamine

ethyl 3-(5-chloropyridin-2-yl)-3-oxopropanoate

  • MDL number MFCD23855581
  • Cas Number : 1106953-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64...66
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137407

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137407
Availability details for
Quantity
  • Availability
  • Add to Cart