ENA413165411 Enamine

1,3-dinitro-1H-pyrazole

  • MDL number MFCD00297297
  • Cas Number : 38858-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68...70
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137799

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-137799
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart