ENA413165418 Enamine

{5-[(tert-butoxy)methyl]-1,2-oxazol-3-yl}methanol

  • MDL number MFCD26935993
  • Cas Number : 1797174-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137850
Availability details for
Quantity
  • Availability
  • Add to Cart