ENA413165419 Enamine

2-bromo-5-fluoropyridin-4-amine

  • MDL number MFCD00234034
  • Cas Number : 887570-94-7
  • Molecular Weight : 191

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-109045

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109045
Availability details for
Quantity
  • Availability
  • Add to Cart