ENA413165421 Enamine

7-chloro-1,4-dihydro-1,6-naphthyridin-4-one

  • MDL number MFCD11044712
  • Cas Number : 952138-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109355

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109355
Availability details for
Quantity
  • Availability
  • Add to Cart