ENA413165423 Enamine

3-(prop-1-en-2-yl)pyridine

  • MDL number MFCD13175169
  • Cas Number : 15825-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137897
Availability details for
Quantity
  • Availability
  • Add to Cart