ENA413165425 Enamine

[5-(4-chlorophenyl)-4-methyl-4H-1,2,4-triazol-3-yl]methanol

  • MDL number MFCD11844957
  • Cas Number : 1126635-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137908

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137908
Availability details for
Quantity
  • Availability
  • Add to Cart