ENA413165435 Enamine

5-methyl-2-(trifluoromethyl)furan-3-carboxylic acid

  • MDL number MFCD00275563
  • Cas Number : 17515-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-109578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109578
Availability details for
Quantity
  • Availability
  • Add to Cart