ENA413165439 Enamine

tert-butyl N-{[4-(hydroxymethyl)-1,3-thiazol-2-yl]methyl}carbamate

  • MDL number MFCD20924985
  • Cas Number : 165667-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138036

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138036
Availability details for
Quantity
  • Availability
  • Add to Cart