ENA413165441 Enamine

5-(3-chloro-4-methoxyphenyl)imidazolidine-2,4-dione

  • MDL number MFCD21150733
  • Cas Number : 1490446-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138056

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138056
Availability details for
Quantity
  • Availability
  • Add to Cart