ENA413165445 Enamine

2-(benzylamino)-1-[3-(difluoromethoxy)phenyl]ethan-1-ol

  • MDL number MFCD26936000
  • Cas Number : 1797802-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95...97
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138074

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138074
Availability details for
Quantity
  • Availability
  • Add to Cart