ENA413165451 Enamine

5,7-dichloro-1,6-naphthyridin-4-ol

  • MDL number MFCD18837806
  • Cas Number : 1443378-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138115
Availability details for
Quantity
  • Availability
  • Add to Cart