ENA413165461 Enamine

methyl 3-phenyl-4,5-dihydro-1,2-oxazole-5-carboxylate

  • MDL number MFCD24741906
  • Cas Number : 17647-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138200

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138200
Availability details for
Quantity
  • Availability
  • Add to Cart