ENA413165463 Enamine

ethyl 2-{1,4-dioxaspiro[4.5]decan-8-ylidene}acetate

  • MDL number MFCD07779434
  • Cas Number : 51656-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-100965

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100965
Availability details for
Quantity
  • Availability
  • Add to Cart