ENA413165467 Enamine

{6-[(2-fluorophenyl)methoxy]pyridin-3-yl}methanol

  • MDL number MFCD19395237
  • Cas Number : 438049-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138239
Availability details for
Quantity
  • Availability
  • Add to Cart