ENA413165477 Enamine

dimethyl[(6-methyl-1H-indol-3-yl)methyl]amine

  • MDL number MFCD01074515
  • Cas Number : 35998-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138333
Availability details for
Quantity
  • Availability
  • Add to Cart