ENA413165478 Enamine

2-(6-chloro-1-methyl-1H-indol-3-yl)acetonitrile

  • MDL number MFCD21986082
  • Cas Number : 1369050-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65...67
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138334

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138334
Availability details for
Quantity
  • Availability
  • Add to Cart