ENA413165481 Enamine

N-(2-bromo-5-methoxyphenyl)-3-phenylprop-2-enamide

  • MDL number MFCD26936009
  • Cas Number : 1562639-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138355

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138355
Availability details for
Quantity
  • Availability
  • Add to Cart