ENA413165492 Enamine

2-(pyrrolidin-1-yl)pyridine

  • MDL number MFCD22083294
  • Cas Number : 54660-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138451

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138451
Availability details for
Quantity
  • Availability
  • Add to Cart