ENA413165493 Enamine

(cyclohexanesulfonyl)benzene

  • MDL number MFCD00091604
  • Cas Number : 6947-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138467

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138467
Availability details for
Quantity
  • Availability
  • Add to Cart