ENA413165496 Enamine

4-phenyl-1,2-oxazole

  • MDL number MFCD01110614
  • Cas Number : 2439-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138480

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138480
Availability details for
Quantity
  • Availability
  • Add to Cart