ENA413165498 Enamine

2-phenyl-2,3,4,5-tetrahydro-1,2,4-triazine-3,5-dione

  • MDL number MFCD18324854
  • Cas Number : 4315-66-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138494

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138494
Availability details for
Quantity
  • Availability
  • Add to Cart