ENA413165502 Enamine

2-phenyl-2,3-dihydropyridazin-3-one

  • MDL number MFCD00234622
  • Cas Number : 14135-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138501

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138501
Availability details for
Quantity
  • Availability
  • Add to Cart