ENA413165506 Enamine

(2-aminoethyl)(2,2-difluoroethyl)methylamine

  • MDL number MFCD12768623
  • Cas Number : 795299-76-2
  • Molecular Weight : 138.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N~1~-(2,2-difluoroethyl)-N~1~-methyl-1,2-ethanediamine
Inchi Code 1S/C5H12F2N2/c1-9(3-2-8)4-5(6)7/h5H,2-4,8H2,1H3
Inchi Key HZWVDKDSDLHMCO-UHFFFAOYSA-N

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-138601

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-138601
pictograms GHS02,GHS05,GHS07
hstatements H226,H302,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart