ENA413165511 Enamine

4-chloro-2-methyl-6-nitrophenol

  • MDL number MFCD09910242
  • Cas Number : 1760-71-0
  • Molecular Weight : 187.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-138642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138642
Availability details for
Quantity
  • Availability
  • Add to Cart