ENA413165526 Enamine

pyrido[2,3-d]pyrimidine-2,4-diol

  • MDL number MFCD07644628
  • Cas Number : 21038-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-138755

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138755
Availability details for
Quantity
  • Availability
  • Add to Cart