ENA413165527 Enamine

4-cyclopropoxybenzene-1-sulfonyl chloride

  • MDL number MFCD12404963
  • Cas Number : 1208075-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138774

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138774
Availability details for
Quantity
  • Availability
  • Add to Cart