ENA413165529 Enamine

methyl 2-[4-(trifluoromethyl)phenyl]-1H-imidazole-4-carboxylate

  • MDL number MFCD17047163
  • Cas Number : 762286-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138781

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138781
Availability details for
Quantity
  • Availability
  • Add to Cart