ENA413165531 Enamine

tert-butyl N-(4-methylthiophen-3-yl)carbamate

  • MDL number MFCD24471163
  • Cas Number : 1097629-78-1
  • Molecular Weight : 213.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (4-methylthiophen-3-yl)carbamate
Inchi Code 1S/C10H15NO2S/c1-7-5-14-6-8(7)11-9(12)13-10(2,3)4/h5-6H,1-4H3,(H,11,12)
Inchi Key XUODWYMBCQAKLB-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-138784

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138784
Availability details for
Quantity
  • Availability
  • Add to Cart