ENA413165542 Enamine

7H-purine-8-thiol

  • MDL number MFCD00022638
  • Cas Number : 583-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138892

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138892
Availability details for
Quantity
  • Availability
  • Add to Cart