ENA413165543 Enamine

methyl 2-(1-methyl-1H-pyrazol-4-yl)acetate

  • MDL number MFCD17267695
  • Cas Number : 1248548-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138905

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-138905
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart