ENA413165544 Enamine

N-[1-(piperidin-4-yl)ethyl]acetamide

  • MDL number MFCD12190947
  • Cas Number : 1156253-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121...123
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138915

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138915
Availability details for
Quantity
  • Availability
  • Add to Cart