ENA413165550 Enamine

3-(6-methylpyridin-3-yl)propan-1-ol

  • MDL number MFCD18802577
  • Cas Number : 50542-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138968

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138968
Availability details for
Quantity
  • Availability
  • Add to Cart