ENA413165563 Enamine

2,2-dimethylcyclopentane-1-carboxylic acid

  • MDL number MFCD11655620
  • Cas Number : 63861-26-7
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,2-dimethylcyclopentanecarboxylic acid
Inchi Code 1S/C8H14O2/c1-8(2)5-3-4-6(8)7(9)10/h6H,3-5H2,1-2H3,(H,9,10)
Inchi Key MQZBBUDJCMHPIP-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-111160

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-111160
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart