ENA413165570 Enamine

2-(4-methyl-3-oxopiperazin-1-yl)acetonitrile

  • MDL number MFCD20389927
  • Cas Number : 1339077-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139109

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139109
Availability details for
Quantity
  • Availability
  • Add to Cart