ENA413165572 Enamine

1-(chloromethyl)-4-fluoro-2-methylbenzene

  • MDL number MFCD11846575
  • Cas Number : 80141-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139114

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139114
Availability details for
Quantity
  • Availability
  • Add to Cart