ENA413165576 Enamine

methyl 8-oxobicyclo[3.2.1]octane-3-carboxylate

  • MDL number MFCD09837132
  • Cas Number : 95685-34-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49...51
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111178

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-111178
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart