ENA413165577 Enamine

ethyl 5-(2-methylcyclopropyl)-1,2,4-oxadiazole-3-carboxylate

  • MDL number MFCD18344019
  • Cas Number : 1339818-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139140

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139140
Availability details for
Quantity
  • Availability
  • Add to Cart