ENA413165590 Enamine

4-benzyl-1,3-benzothiazol-2-amine

  • MDL number MFCD10693533
  • Cas Number : 1152504-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132...134
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139196

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139196
Availability details for
Quantity
  • Availability
  • Add to Cart