ENA413165607 Enamine

(4-cyclobutylphenyl)boronic acid

  • MDL number MFCD12760760
  • Cas Number : 845797-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163...165
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139273

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139273
Availability details for
Quantity
  • Availability
  • Add to Cart