ENA413165615 Enamine

2-cyano-2,2-dimethylacetamide

  • MDL number MFCD16105926
  • Cas Number : 7505-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-106
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139359
Availability details for
Quantity
  • Availability
  • Add to Cart