ENA413165619 Enamine

N-(2,2,2-trifluoroethyl)-1H-1,3-benzodiazole-5-carboxamide

  • MDL number MFCD11727439
  • Cas Number : 1110885-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139408
Availability details for
Quantity
  • Availability
  • Add to Cart