ENA413165626 Enamine

1-methoxycyclohexane-1-carboxylic acid

  • MDL number MFCD14635185
  • Cas Number : 50421-27-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139440

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139440
Availability details for
Quantity
  • Availability
  • Add to Cart