ENA413165827 Enamine

3-methyl-1-(pyridin-2-ylmethyl)piperazine dihydrochloride

  • MDL number MFCD26936050
  • Cas Number : 1803580-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139647

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139647
Availability details for
Quantity
  • Availability
  • Add to Cart