ENA413165830 Enamine

ethyl 6-(methylsulfamoyl)pyridine-3-carboxylate

  • MDL number MFCD26936052
  • Cas Number : 1803566-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66...68
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139812

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139812
Availability details for
Quantity
  • Availability
  • Add to Cart