ENA413165831 Enamine

5-(2-methoxyethoxy)-1,3-dimethyl-1H-pyrazole

  • MDL number MFCD26936053
  • Cas Number : 1803606-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139835

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139835
Availability details for
Quantity
  • Availability
  • Add to Cart