ENA413165833 Enamine

methyl 5-(benzylsulfanyl)-3-methyl-1H-pyrazole-4-carboxylate

  • MDL number MFCD26936055
  • Cas Number : 73743-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139880

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139880
Availability details for
Quantity
  • Availability
  • Add to Cart